Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163159.1 | NP_001156631.1 | 4 | 596 | ||
NM_001163160.1 | NP_001156632.1 | 4 | 530 | |||
NM_009981.4 | NP_034111.1 | 4 | 432 | |||
GenBank mRNA | AK044585.1 | - | 4 | 433 | ||
AK076050.1 | - | 4 | 357 | |||
AK076830.1 | - | 4 | 531 | |||
BB642590.1 | - | 4 | 433 | |||
BC018313.1 | - | 3 | 315 | 4162053 | ||
L28956.1 | - | 4 | 401 | |||
Z12302.1 | - | 4 | 391 |