Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000519.3 | NP_000510.1 | 1 | 256 | ||
GenBank mRNA | AY034468.1 | - | 1 | 256 | ||
BC069307.1 | - | 1 | 111 | 7262103 | ||
BC070282.1 | - | 1 | 112 | 30418964 | ||
BU664913.1 | - | 1 | 75 |