Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010011.3 | NP_034141.3 | 7 | 837 | ||
GenBank mRNA | AB017785.1 | - | 7 | 797 | ||
AB018421.1 | - | 7 | 831 | |||
AK002528.1 | - | 7 | 838 | |||
AK098101.1 | - | 7 | 835 | |||
BC010747.1 | - | 7 | 822 | 4242518 | ||
BC051049.1 | - | 7 | 823 | 4234581 | ||
X69296.1 | - | 811 |