Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4a12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172306.2 | NP_758510.2 | 8 | 968 | ||
GenBank mRNA | AK098123.1 | - | 8 | 949 | ||
BC060945.1 | - | 8 | 949 | 30304458 |
Gene Symbol: Cyp4a12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177406.3 | NP_803125.2 | 8 | 991 | ||
GenBank mRNA | AK144260.1 | - | 8 | 921 | ||
BC014721.1 | - | 8 | 937 | 4240359 | ||
BC025936.1 | - | 8 | 931 | 5097698 | ||
BC026582.1 | - | 8 | 937 | 4971335 | ||
BC031141.1 | - | 8 | 912 | 4972684 | ||
BC033924.1 | - | 8 | 921 | 5101821 |