Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010014.3 | NP_034144.1 | 5 | 693 | ||
NM_177259.4 | NP_796233.2 | 5 | 693 | |||
GenBank mRNA | AK005640.1 | - | 4 | 504 | ||
BC051993.1 | - | 7 | 826 | |||
BC138538.1 | - | 4 | 414 | 8861558 | ||
BC138539.1 | - | 4 | 414 | 8861559 | ||
Y08379.1 | - | 5 | 641 | |||
Y08380.1 | - | 5 | 641 | |||
Y08381.1 | - | 5 | 641 |