Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190451.1 | NP_001177380.1 | 3, 4 | 633 | ||
NM_007833.5 | NP_031859.1 | 3, 4 | 541 | |||
GenBank mRNA | AK135287.1 | - | 3, 4 | 525 | ||
AK145905.1 | - | 3, 4 | 523 | |||
AK147020.1 | - | 3, 4 | 525 | |||
AK161788.1 | - | 3, 4 | 523 | |||
AK165408.1 | - | 3, 4 | 523 | |||
BC060126.1 | - | 2, 3 | 354 | |||
BC132521.1 | - | 3, 4 | 392 | 40130798 | ||
BC138564.1 | - | 3, 4 | 392 | 8861584 | ||
X53929.1 | - | 3, 4 | 550 | |||
Y11757.1 | - | 2, 3 | 239 |