Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110222.1 | NP_001103692.1 | 3 | 845 | ||
NM_001110223.1 | NP_001103693.1 | 3 | 767 | |||
NM_001110224.1 | NP_001103694.1 | 3 | 767 | |||
NM_010025.2 | NP_034155.2 | 3 | 767 | |||
GenBank mRNA | AB011678.1 | - | 3 | 863 | ||
AF045547.1 | - | 3 | 648 | |||
AK014264.1 | - | 3 | 768 | |||
AK048410.1 | - | 3 | 773 | |||
AK049050.1 | - | 3 | 841 | |||
AK164119.1 | - | 3 | 654 | |||
AY560329.1 | - | 3 | 655 | |||
BC056391.1 | - | 3 | 660 | 6855006 | ||
BC057010.1 | - | 3 | 752 | 6825750 | ||
BC062974.1 | - | 3 | 752 | 6825750 |