Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007852.1 | NP_031878.1 | 2 | 194 | ||
GenBank mRNA | U03034.1 | - | 1 | 22 |
Gene Symbol: Defa12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U03063.1 | - | 1 | 22 |
Gene Symbol: Defa24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024225.2 | NP_001019396.1 | 2 | 237 | ||
GenBank mRNA | AY746429.1 | - | 2 | 194 |
Gene Symbol: Gm15284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177485.1 | NP_001170956.1 | 2 | 207 |