Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010046.2 | NP_034176.1 | 4, 5 | 476 | ||
GenBank mRNA | AB057816.1 | - | 4, 5 | 464 | ||
AF078752.1 | - | 4, 5 | 534 | |||
BC003717.1 | - | 4, 5 | 476 | 3486724 |