Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010049.3 | NP_034179.1 | 5 | 882 | ||
GenBank mRNA | AK031241.1 | - | 5 | 516 | ||
AK136212.1 | - | 5 | 940 | |||
BC005796.1 | - | 5 | 454 | 3707582 | ||
V00734.1 | - | 5 | 518 | |||
X56066.1 | - | 5 | 428 |