Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010062.3 | NP_034192.1 | 5 | 682 | ||
GenBank mRNA | AK145192.1 | - | 5 | 683 | ||
AK167277.1 | - | 5 | 648 | |||
BC058609.1 | - | 5 | 629 | 5397016 |