Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1i2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198872.1 | NP_001185801.1 | 3 | 354 | ||
NM_001198873.1 | NP_001185802.1 | 3 | 354 | |||
NM_001198874.1 | NP_001185803.1 | 3 | 354 | |||
NM_001198875.1 | NP_001185804.1 | 3 | 354 | |||
NM_001198876.1 | NP_001185805.1 | 3 | 354 | |||
NM_001198877.1 | NP_001185806.1 | 3 | 343 | |||
NM_001198878.1 | NP_001185807.1 | 3 | 343 | |||
NM_010064.4 | NP_034194.1 | 3 | 343 | |||
GenBank mRNA | AF063231.1 | - | 3 | 282 | ||
AK088015.1 | - | 3 | 314 | |||
AK135477.1 | - | 3 | 285 | |||
AK141837.1 | - | 3 | 322 | |||
AK150173.1 | - | 3 | 304 | |||
AK164322.1 | - | 3 | 278 | |||
AK164437.1 | - | 3 | 278 | |||
AK168730.1 | - | 3 | 318 | |||
BC139374.1 | - | 3 | 238 | 8862396 | ||
BC139375.1 | - | 3 | 238 | 8862397 | ||
BY065048.1 | - | 3 | 357 | |||
BY226221.1 | - | 3 | 344 | |||
GU992208.1 | - | 3 | 245 | |||
GU992209.1 | - | 3 | 245 | |||
GU992210.1 | - | 3 | 245 | |||
GU992211.1 | - | 3 | 283 | |||
GU992212.1 | - | 3 | 283 |