Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291799.1 | NP_001278728.1 | 5 | 832 | ||
NM_010070.4 | NP_034200.4 | 5 | 834 | |||
GenBank mRNA | AA790529.1 | - | 3 | 369 | ||
AK136382.1 | - | 5 | 835 | |||
AK169871.1 | - | 4 | 1000 | |||
BC013066.1 | - | 5 | 828 | 3498170 | ||
U78818.2 | - | 5 | 799 |