Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291799.1 | NP_001278728.1 | 2, 3 | 391 | ||
NM_010070.4 | NP_034200.4 | 2, 3 | 393 | |||
GenBank mRNA | AK136382.1 | - | 2, 3 | 394 | ||
AK169871.1 | - | 2, 3 | 389 | |||
BC013066.1 | - | 2, 3 | 387 | 3498170 | ||
CJ053718.1 | - | 2, 3 | 394 | |||
U78818.2 | - | 2, 3 | 358 |