Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007878.2 | NP_031904.1 | 4 | 1280 | ||
GenBank mRNA | AK044379.1 | - | 4 | 1249 | ||
AK044436.1 | - | 4 | 1271 | |||
BC016086.1 | - | 4 | 1280 | 4501819 | ||
BC051421.1 | - | 4 | 1256 | 6492837 |
Set Membership: |
G-protein coupled receptor |