Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsg1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010079.2 | NP_034209.2 | 15 | 3884 | ||
GenBank mRNA | AK029294.1 | - | 1 | 1541 | ||
AK037234.1 | - | 1 | 757 |