Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007883.2 | NP_031909.2 | 4 | 533 | ||
GenBank mRNA | AB072269.1 | - | 4 | 561 | ||
BC020144.1 | - | 4 | 427 | |||
BC044848.1 | - | 4 | 494 | |||
BC131653.1 | - | 4 | 509 | 40110529 | ||
BC131654.1 | - | 4 | 509 | 40110531 |