Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113414.1 | NP_001106885.1 | 14 | 1973 | ||
NM_001113415.1 | NP_001106886.1 | 14 | 2000 | |||
NM_010096.3 | NP_034226.1 | 14 | 1973 | |||
GenBank mRNA | AK138602.1 | - | 9 | 1162 | ||
AK220542.1 | - | 14 | 1973 | |||
BC064016.1 | - | 4 | 374 | |||
BC067018.1 | - | 15 | 1728 | 6833120 | ||
U92702.1 | - | 14 | 1541 | |||
U92704.1 | - | 14 | 1514 |