Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252653.1 | NP_001239582.1 | 8 | 915 | ||
NM_007899.2 | NP_031925.2 | 8 | 918 | |||
GenBank mRNA | AK153009.1 | - | 8 | 917 | ||
AK153471.1 | - | 8 | 917 | |||
AK159334.1 | - | 8 | 914 | |||
AK169939.1 | - | 8 | 920 | |||
BC138693.1 | - | 8 | 904 | 8861715 | ||
BC145381.1 | - | 8 | 901 | 9053939 | ||
L33416.1 | - | 8 | 911 |