Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000187.3 | NP_000178.2 | 9 | 1034 | ||
XM_005247413.1 | XP_005247470.1 | 9 | 1034 | |||
XM_005247414.4 | XP_005247471.1 | 9 | 1034 | |||
XM_011512746.1 | XP_011511048.1 | 9 | 1034 | |||
XM_017006277.1 | XP_016861766.1 | 9 | 647 | |||
GenBank mRNA | AF045167.1 | - | 9 | 741 | ||
AK124291.1 | - | 4 | 1418 | |||
AK290782.1 | - | 9 | 740 | |||
AK313563.1 | - | 9 | 1018 | |||
BC020792.1 | - | 9 | 740 | 4716316 | ||
BC071757.1 | - | 9 | 672 | 6714217 | ||
U63008.1 | - | 9 | 703 | |||
Z75048.1 | - | 9 | 703 |