Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010111.5 | NP_034241.2 | 1 | 250 | ||
GenBank mRNA | AK132512.1 | - | 1 | 717 | ||
AK132845.1 | - | 1 | 543 | |||
AK134108.1 | - | 1 | 717 | |||
AK142203.1 | - | 1 | 720 | |||
AK142283.1 | - | 1 | 720 | |||
AK143059.1 | - | 1 | 717 | |||
BC057009.1 | - | 1 | 538 | 6827408 | ||
CT010381.1 | - | 1 | 91 | |||
L38847.1 | - | 1 | 251 | |||
U16819.1 | - | 1 | 119 | |||
U30244.1 | - | 1 | 88 |