Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ei24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199494.1 | NP_001186423.1 | 9 | 947 | ||
NM_007915.5 | NP_031941.1 | 9 | 831 | |||
GenBank mRNA | AK145018.1 | - | 9 | 908 | ||
AK147776.1 | - | 9 | 860 | |||
AK159927.1 | - | 9 | 889 | |||
AK172233.1 | - | 9 | 911 | |||
U41751.2 | - | 9 | 835 |