Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010123.3 | NP_034253.3 | 16 | 2585 | ||
GenBank mRNA | AK013070.1 | - | 6 | 900 | ||
AK145629.1 | - | 16 | 2550 | |||
AK162784.1 | - | 5 | 498 | |||
AK172899.1 | - | 16 | 2522 | |||
U14172.1 | - | 16 | 2535 | |||
X17373.1 | - | 9 | 1204 | |||
X84651.1 | - | 9 | 1205 |