Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013515.2 | NP_038543.1 | 2 | 211 | ||
GenBank mRNA | AF093620.1 | - | 2 | 162 | ||
AK077435.1 | - | 2 | 179 | |||
AK139242.1 | - | 2 | 179 | |||
AK143724.1 | - | 2 | 179 | |||
AK148975.1 | - | 2 | 180 | |||
AK149689.1 | - | 2 | 179 | |||
AK149821.1 | - | 2 | 179 | |||
AK149991.1 | - | 2 | 179 | |||
AK151129.1 | - | 2 | 178 | |||
AK181815.1 | - | 2 | 223 | |||
BC003789.1 | - | 2 | 174 | 3593052 | ||
U17297.1 | - | 2 | 211 |