Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000188.2 | NP_000179.2 | 17, 18 | 2711 | ||
NM_001322364.1 | NP_001309293.1 | 19, 20 | 2804 | |||
NM_001322365.1 | NP_001309294.1 | 22, 23 | 3109 | |||
NM_001322366.1 | NP_001309295.1 | 17, 18 | 2602 | |||
NM_001322367.1 | NP_001309296.1 | 16, 17 | 2615 | |||
NM_033496.2 | NP_277031.1 | 17, 18 | 2708 | |||
NM_033497.2 | NP_277032.1 | 20, 21 | 2926 | |||
NM_033498.2 | NP_277033.1 | 20, 21 | 2980 | |||
NM_033500.2 | NP_277035.2 | 21, 22 | 3073 | |||
XM_005269737.1 | XP_005269794.1 | 17, 18 | 2982 | |||
XM_011539732.1 | XP_011538034.1 | 21, 22 | 2980 | |||
GenBank mRNA | AB209526.1 | - | 17, 18 | 2707 | ||
AK128226.1 | - | 22, 23 | 3104 | |||
AK292012.1 | - | 17, 18 | 2711 | |||
AK294428.1 | - | 17, 18 | 2708 | |||
BC008730.2 | - | 17, 18 | 2666 | 3163058 | ||
M75126.1 | - | 17, 18 | 2691 | |||
X66957.1 | - | 11, 12 | 1752 |