Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271587.1 | NP_001258516.1 | 7 | 877 | ||
NM_001271588.1 | NP_001258517.1 | 7 | 808 | |||
NM_001271595.1 | NP_001258524.1 | 7 | 882 | |||
NM_007945.3 | NP_031971.2 | 8 | 883 | |||
GenBank mRNA | AK147877.1 | - | 8 | 811 | ||
AK148424.1 | - | 8 | 799 | |||
AK149683.1 | - | 7 | 671 | |||
AK155022.1 | - | 7 | 643 | |||
AK166156.1 | - | 8 | 718 | |||
BC016890.1 | - | 7 | 894 | 4240899 | ||
L21671.1 | - | 7 | 798 |