Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271587.1 | NP_001258516.1 | 15, 16 | 1886 | ||
NM_001271588.1 | NP_001258517.1 | 15, 16 | 1817 | |||
NM_001271589.1 | NP_001258518.1 | 8, 9 | 887 | |||
NM_001271595.1 | NP_001258524.1 | 15, 16 | 1891 | |||
NM_007945.3 | NP_031971.2 | 16, 17 | 1892 | |||
GenBank mRNA | AK136246.1 | - | 6, 7 | 658 | ||
AK147877.1 | - | 16, 17 | 1820 | |||
AK149683.1 | - | 15, 16 | 1680 | |||
AK155022.1 | - | 15, 16 | 1652 | |||
AK165684.1 | - | 8, 9 | 890 | |||
AK166156.1 | - | 16, 17 | 1727 | |||
AK187916.1 | - | 3, 4 | 364 | |||
BC016890.1 | - | 15, 16 | 1903 | 4240899 | ||
L21671.1 | - | 15, 16 | 1807 |