Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002115.2 | NP_002106.2 | 8 | 972 | ||
XM_011534540.2 | XP_011532842.1 | 8 | 1013 | |||
XM_017009411.1 | XP_016864900.1 | 7 | 1188 | |||
GenBank mRNA | AB208951.1 | - | 8 | 1171 | ||
BC028129.1 | - | 8 | 972 | 5211860 | ||
U51333.1 | - | 8 | 954 |