Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252193.1 | NP_001239122.1 | 8 | 1440 | ||
NM_025275.4 | NP_079551.3 | 9 | 1259 | |||
GenBank mRNA | AJ879914.1 | - | 6 | 956 | ||
AK004062.1 | - | 9 | 1219 | |||
AK075748.1 | - | 9 | 1199 | |||
BC025087.1 | - | 8 | 1434 | 2649126 |