Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024166.6 | NP_077128.2 | 4 | 804 | ||
GenBank mRNA | AK003399.1 | - | 4 | 639 | ||
AK012691.1 | - | 3 | 630 | |||
AK088374.1 | - | 4 | 805 | |||
AK145492.1 | - | 4 | 622 | |||
AK160762.1 | - | 4 | 623 | |||
AK160779.1 | - | 4 | 624 | |||
AK162133.1 | - | 4 | 622 | |||
AK166796.1 | - | 4 | 624 | |||
AK167593.1 | - | 4 | 624 | |||
AK210686.1 | - | 2 | 152 | |||
AK211922.1 | - | 1 | 153 | |||
AK216042.1 | - | 2 | 152 | |||
BC037682.1 | - | 3 | 1984 | |||
BC042717.1 | - | 4 | 611 | 3154544 | ||
BC069887.1 | - | 4 | 386 | |||
BC082287.1 | - | 4 | 614 | 6397957 | ||
BC083126.1 | - | 3 | 1947 | |||
BC084682.1 | - | 4 | 593 | 6530688 |