Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163154.1 | NP_001156626.1 | 6 | 740 | ||
NM_007960.4 | NP_031986.1 | 9 | 1080 | |||
GenBank mRNA | AK032224.1 | - | 6 | 742 | ||
AK035914.1 | - | 7 | 1471 | |||
AK144713.1 | - | 9 | 1029 | |||
BC005645.1 | - | 9 | 1008 | 3257346 | ||
CK619331.1 | - | 2 | 154 | |||
L10426.1 | - | 9 | 1029 |