Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161373.1 | NP_001154845.1 | 3 | 725 | ||
NM_001161374.1 | NP_001154846.1 | 3 | 725 | |||
NM_007977.2 | NP_032003.2 | 3 | 725 | |||
GenBank mRNA | AJ627040.1 | - | 2 | 145 | ||
AK051579.1 | - | 3 | 3001 | |||
BC138347.1 | - | 3 | 606 | 8861367 | ||
BC145245.1 | - | 3 | 606 | 9053797 | ||
L05573.1 | - | 3 | 723 |