Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081437.1 | NP_001074906.1 | 2 | 1353 | ||
NM_007992.2 | NP_032018.2 | 2 | 1353 | |||
GenBank mRNA | AK159361.1 | - | 2 | 1665 | ||
AK161522.1 | - | 2 | 1296 | |||
AK168688.1 | - | 2 | 1291 | |||
BC005443.1 | - | 2 | 1333 | 3490759 | ||
X75285.1 | - | 2 | 1253 |