Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010188.5 | NP_034318.2 | 5 | 701 | ||
GenBank mRNA | AF197930.1 | - | 5 | 696 | ||
AK077227.1 | - | 5 | 661 | |||
AK151557.1 | - | 5 | 661 | |||
AK152875.1 | - | 5 | 661 | |||
AK154892.1 | - | 5 | 662 | |||
AK163401.1 | - | 3 | 651 | |||
AK169887.1 | - | 5 | 658 | |||
AK171289.1 | - | 5 | 662 | |||
AY897419.1 | - | 5 | 650 | |||
AY897420.1 | - | 5 | 646 | |||
AY897421.1 | - | 5 | 650 | |||
AY897422.1 | - | 5 | 650 | |||
AY897423.1 | - | 5 | 650 | |||
AY897424.1 | - | 5 | 645 | |||
AY897425.1 | - | 5 | 655 | |||
AY897426.1 | - | 5 | 655 | |||
AY897427.1 | - | 5 | 655 | |||
AY897428.1 | - | 5 | 659 | |||
AY897429.1 | - | 5 | 650 | |||
M14215.1 | - | 5 | 696 |