Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010189.3 | NP_034319.2 | 5 | 1205 | ||
GenBank mRNA | AK146933.1 | - | 5 | 1170 | ||
AK152879.1 | - | 5 | 1141 | |||
BC003786.1 | - | 5 | 1140 | 3485649 | ||
D37874.1 | - | 5 | 1138 | |||
L17022.1 | - | 4 | 844 |
Gene Symbol: Mup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC093515.1 | - | 2402 |