Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007995.3 | NP_032021.1 | 10 | 1058 | ||
GenBank mRNA | AB007813.1 | - | 10 | 950 | ||
AB222271.1 | - | 9 | 839 | |||
AK133539.1 | - | 10 | 1035 | |||
AK138752.1 | - | 10 | 1005 | |||
AK159651.1 | - | 10 | 1028 | |||
BC019180.1 | - | 10 | 1006 | 5008934 | ||
CT010383.1 | - | 10 | 920 |