Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il4i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010215.3 | NP_034345.2 | 3 | 172 | ||
GenBank mRNA | AK150582.1 | - | 3 | 169 | ||
AK151030.1 | - | 3 | 169 | |||
AK151171.1 | - | 3 | 169 | |||
AK151731.1 | - | 3 | 173 | |||
AK152538.1 | - | 3 | 169 | |||
AK157036.1 | - | 2 | 358 | |||
AY157538.1 | - | 3 | 173 | |||
BC115960.1 | - | 3 | 160 | 40054305 | ||
U70429.1 | - | 3 | 172 |
Gene Symbol: Nup62-il4i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171024.1 | NP_001164495.1 | 4 | 431 | ||
GenBank mRNA | AK133082.1 | - | 4 | 406 | ||
CJ047381.1 | - | 4 | 406 |