Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111066.1 | NP_001104536.1 | 5, 6 | 865 | ||
NM_001199631.1 | NP_001186560.1 | 5, 6 | 961 | |||
NM_010223.2 | NP_034353.2 | 5, 6 | 862 | |||
GenBank mRNA | AB154247.1 | - | 4, 5 | 601 | ||
AF030635.1 | - | 4, 5 | 632 | |||
AK088739.1 | - | 5, 6 | 800 | |||
AK146122.1 | - | 5, 6 | 860 | |||
AK152198.1 | - | 5, 6 | 861 | |||
AK153000.1 | - | 5, 6 | 858 | |||
AK165281.1 | - | 5, 6 | 858 | |||
AK167663.1 | - | 5, 6 | 848 | |||
AK177591.1 | - | 1, 2 | 56 | |||
AK179469.1 | - | 1, 2 | 109 | |||
AK209719.1 | - | 2, 3 | 265 | |||
AK212094.1 | - | 2, 3 | 263 | |||
AY225340.1 | - | 4, 5 | 745 | |||
AY278608.1 | - | 5, 6 | 821 | |||
BC003739.1 | - | 4, 5 | 781 | 3601511 | ||
BC027808.1 | - | 5, 6 | 815 | 5324380 |