Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008047.5 | NP_032073.2 | 9, 10 | 890 | ||
GenBank mRNA | AK049440.1 | - | 9, 10 | 891 | ||
AK147117.1 | - | 9, 10 | 888 | |||
BC006185.1 | - | 9, 10 | 918 | |||
BC028921.1 | - | 9, 10 | 874 | 2647002 | ||
M91380.1 | - | 9, 10 | 871 |