Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ganab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293621.1 | NP_001280550.1 | 10 | 1204 | ||
NM_008060.2 | NP_032086.1 | 11 | 1270 | |||
GenBank mRNA | AK028655.1 | - | 10 | 1174 | ||
AK030722.1 | - | 10 | 1173 | |||
AK047454.1 | - | 5 | 525 | |||
AK081915.1 | - | 10 | 1166 | |||
AK122201.1 | - | 5 | 1400 | |||
AK149686.1 | - | 11 | 1227 | |||
AK166150.1 | - | 10 | 1143 | |||
BC020386.1 | - | 3 | 420 | |||
BC035423.1 | - | 1 | 20 | |||
BC094437.1 | - | 10 | 1154 | 30682580 | ||
BC117888.1 | - | 10 | 1147 | 40096413 | ||
BC117889.1 | - | 10 | 1150 | 40096414 | ||
BC128069.1 | - | 11 | 1214 | 40096428 | ||
U92793.1 | - | 11 | 1216 |