Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194999.1 | NP_001181928.1 | 4 | 545 | ||
NM_001195000.1 | NP_001181929.1 | 4 | 680 | |||
NM_001195035.1 | NP_001181964.1 | 4 | 680 | |||
NM_001310213.1 | NP_001297142.1 | 5 | 796 | |||
NM_001531.2 | NP_001522.1 | 4 | 680 | |||
XM_006711286.3 | XP_006711349.1 | 4 | 726 | |||
XM_011509463.2 | XP_011507765.1 | 3 | 452 | |||
GenBank mRNA | AF010446.1 | - | 3 | 524 | ||
AF010447.1 | - | 3 | 505 | |||
AF031469.1 | - | 3 | 507 | |||
AJ249778.1 | - | 3 | 524 | |||
AK222910.1 | - | 3 | 533 | |||
AK290374.1 | - | 3 | 544 | |||
AK304645.1 | - | 3 | 596 | |||
BC012485.1 | - | 3 | 389 | 4540305 | ||
BX415555.2 | - | 4 | 632 | |||
U22963.1 | - | 3 | 524 |