Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000411.6 | NP_000402.3 | 11 | 2298 | ||
NM_001242784.1 | NP_001229713.1 | 11 | 3232 | |||
NM_001242785.1 | NP_001229714.1 | 11 | 2391 | |||
XM_005260953.3 | XP_005261010.1 | 10 | 2260 | |||
XM_005260955.3 | XP_005261012.1 | 10 | 2131 | |||
XM_005260956.3 | XP_005261013.1 | 10 | 2307 | |||
XM_006723994.2 | XP_006724057.1 | 10 | 2245 | |||
XM_006723995.1 | XP_006724058.1 | 11 | 2423 | |||
XM_011529538.1 | XP_011527840.1 | 11 | 2278 | |||
XM_011529539.2 | XP_011527841.1 | 11 | 2283 | |||
XM_011529541.2 | XP_011527843.1 | 10 | 2134 | |||
XM_017028330.1 | XP_016883819.1 | 10 | 2081 | |||
GenBank mRNA | AK314189.1 | - | 10 | 2165 | ||
BC060787.1 | - | 11 | 2256 | 30343493 | ||
D23672.2 | - | 11 | 2289 | |||
D87328.1 | - | 11 | 3049 | |||
X80160.1 | - | 10 | 2149 |