Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008077.4 | NP_032103.2 | 12 | 1402 | ||
GenBank mRNA | AF326547.1 | - | 12 | 1352 | ||
AF326548.1 | - | 12 | 1352 | |||
AF483492.1 | - | 11 | 1169 | |||
AF483493.1 | - | 11 | 1169 | |||
AK136801.1 | - | 12 | 1387 | |||
AK148497.1 | - | 12 | 1402 | |||
AK158827.1 | - | 11 | 1168 | |||
AK160410.1 | - | 11 | 1311 | |||
AK160868.1 | - | 12 | 1400 | |||
BC027059.1 | - | 12 | 1353 | 5358787 | ||
KC134210.1 | - | 11 | 1169 | |||
S67453.1 | - | 7 | 578 | |||
Y12257.1 | - | 11 | 1169 | |||
Z49976.1 | - | 12 | 1289 |