Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008081.3 | NP_032107.1 | 3 | 319 | ||
GenBank mRNA | BC071231.1 | - | 3 | 287 | ||
BC139165.1 | - | 3 | 309 | 8862187 | ||
BC139166.1 | - | 3 | 309 | 8862188 | ||
L30104.1 | - | 3 | 319 |