Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077411.2 | NP_001070879.1 | 7 | 893 | ||
NM_008094.5 | NP_032120.1 | 8 | 1062 | |||
GenBank mRNA | AK082767.1 | - | 7 | 891 | ||
AK152745.1 | - | 7 | 874 | |||
AK166198.1 | - | 8 | 1053 | |||
BC006663.1 | - | 8 | 1047 | 3498551 | ||
M24119.1 | - | 7 | 802 |