Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008105.3 | NP_032131.2 | 3 | 1693 | ||
NM_023887.4 | NP_076376.3 | 3 | 1671 | |||
NM_133219.1 | NP_573482.1 | 3 | 1333 | |||
GenBank mRNA | AB037597.1 | - | 3 | 1084 | ||
AK054226.1 | - | 3 | 1208 | |||
AK077598.1 | - | 3 | 1308 | |||
AK080061.1 | - | 3 | 1260 | |||
AK146638.1 | - | 3 | 435 | |||
AK146679.1 | - | 3 | 435 | |||
AK156328.1 | - | 3 | 1207 | |||
AK169167.1 | - | 3 | 435 | |||
AY236873.1 | - | 3 | 1568 | |||
AY236874.1 | - | 3 | 1471 | |||
AY236875.1 | - | 3 | 1366 | |||
AY435148.1 | - | 3 | 1084 | |||
AY435149.1 | - | 3 | 1084 | |||
AY435150.1 | - | 3 | 1084 | |||
BC054845.1 | - | 3 | 1315 | 6493137 | ||
BC094572.1 | - | 3 | 1671 | 30356538 | ||
BQ892107.1 | - | 3 | 841 | |||
U68182.1 | - | 3 | 1317 |