Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gjb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010937.2 | NP_001010937.1 | 3 | 1097 | ||
NM_001271663.1 | NP_001258592.1 | 3 | 1188 | |||
GenBank mRNA | AK134617.1 | - | 3 | 1096 | ||
AK141610.1 | - | 3 | 1097 | |||
BC013811.1 | - | 3 | 764 | 3483477 | ||
BC016507.1 | - | 3 | 772 | 3481872 | ||
BC063762.1 | - | 1 | 911 | 30245263 |