Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010305.1 | NP_034435.1 | 5 | 720 | ||
GenBank mRNA | BC032995.1 | - | 5 | 662 | ||
BC138862.1 | - | 5 | 600 | 8861884 | ||
BC138865.1 | - | 5 | 600 | 8861887 |