Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025277.3 | NP_079553.1 | 3 | 941 | ||
GenBank mRNA | AK003187.1 | - | 3 | 879 | ||
BC023670.1 | - | 3 | 884 | 5031104 | ||
BC055434.1 | - | 4 | 866 | 3994982 | ||
BC061090.1 | - | 3 | 864 |