Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010320.3 | NP_034450.1 | 3 | 699 | ||
GenBank mRNA | AK144609.1 | - | 3 | 700 | ||
BC111437.1 | - | 1 | 63 | 40049157 |